siRNA Details
| Gene Symbol: Rsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232102.2 | XP_006232164.1 | 3 | 492 | ||
| XM_006232103.2 | XP_006232165.1 | 3 | 761 | |||
| XM_006232104.2 | XP_006232166.1 | 3 | 216 | |||
| XM_006232105.2 | XP_006232167.1 | 3 | 516 | |||
| XM_006232106.2 | XP_006232168.1 | 3 | 224 | |||
| XM_008760847.1 | XP_008759069.1 | 3 | 804 | |||

Rsl1
Genomic Map