siRNA Details
| Gene Symbol: GREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014668.3 | NP_055483.2 | 5 | 816 | ||
| NM_033090.2 | NP_149081.1 | 5 | 767 | |||
| NM_148903.2 | NP_683701.2 | 5 | 832 | |||
| XM_005246192.4 | XP_005246249.1 | 5 | 838 | |||
| XM_011510418.2 | XP_011508720.1 | 4 | 677 | |||
| XM_011510419.2 | XP_011508721.1 | 4 | 677 | |||
| XM_011510423.2 | XP_011508725.1 | 4 | 677 | |||
| GenBank mRNA | AF245388.1 | - | 5 | 816 | ||
| AF245389.1 | - | 5 | 767 | |||
| AF245390.1 | - | 5 | 832 | |||
| BC071853.1 | - | 4 | 497 | |||

GREB1
Genomic Map