siRNA Details
| Gene Symbol: LOC683722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101008.1 | NP_001094478.1 | 5 | 466 | ||
| XM_008758772.1 | XP_008756994.1 | 5 | 877 | |||
| XM_008758773.1 | XP_008756995.1 | 5 | 874 | |||
| XM_008758774.1 | XP_008756996.1 | 5 | 571 | |||
| GenBank mRNA | BC098026.1 | - | 5 | 466 | 7455942 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_201230, NM_001197046, BC080717, BB615739, AK050389, and others.

LOC683722
Genomic Map