siRNA Details
| Gene Symbol: Dydc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252817.2 | XP_006252879.1 | 2, 3 | 548 | ||
| XM_006252818.2 | XP_006252880.1 | 2, 3 | 208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dydc1| Gene Symbol: Dydc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252817.2 | XP_006252879.1 | 2, 3 | 548 | ||
| XM_006252818.2 | XP_006252880.1 | 2, 3 | 208 | |||