siRNA Details
| Gene Symbol: Clec2dl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085404.1 | NP_001078873.1 | 3 | 316 | ||
| XM_006237453.2 | XP_006237515.1 | 4 | 349 | |||
| XM_008763333.1 | XP_008761555.1 | 3 | 1375 | |||
| GenBank mRNA | EF100689.1 | - | 3 | 316 | ||
| FJ404471.1 | - | 3 | 316 | |||

Clec2dl1
Genomic Map