siRNA Details
| Gene Symbol: Cfhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001068704.5 | XP_001068704.3 | 11 | 1933 | ||
| XM_006249947.2 | XP_006250009.1 | 11 | 1859 | |||
| XM_006249948.2 | XP_006250010.1 | 10 | 1763 | |||
| XM_006249949.2 | XP_006250011.1 | 10 | 1756 | |||
| XM_006249950.2 | XP_006250012.1 | 9 | 1573 | |||
| XM_006249951.2 | XP_006250013.1 | 9 | 1568 | |||
| GenBank mRNA | FQ211198.1 | - | 10 | 1736 | ||

Cfhr2
Genomic Map