siRNA Details
| Gene Symbol: UBE3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014671.2 | NP_055486.2 | 7 | 1006 | ||
| XM_005249564.4 | XP_005249621.1 | 6 | 940 | |||
| XM_017012818.1 | XP_016868307.1 | 8 | 1278 | |||
| GenBank mRNA | AK295138.1 | - | 7 | 686 | ||
| BC014029.2 | - | 8 | 766 | 4541634 | ||
| BC026241.1 | - | 6 | 881 | |||
| D13635.1 | - | 7 | 997 | |||

UBE3C
Genomic Map