siRNA Details
| Gene Symbol: Cfhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001068704.5 | XP_001068704.3 | 5 | 865 | ||
| XM_006249947.2 | XP_006250009.1 | 5 | 791 | |||
| XM_006249948.2 | XP_006250010.1 | 5 | 866 | |||
| XM_006249951.2 | XP_006250013.1 | 5 | 866 | |||

Cfhr2
Genomic Map