siRNA Details
| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013053.1 | NP_037185.1 | 3, 4 | 495 | ||
| GenBank mRNA | BC062409.1 | - | 2, 3 | 465 | 5623850 | |
| D17614.1 | - | 3, 4 | 495 | |||
| FQ218679.1 | - | 2, 3 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ywhaq| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013053.1 | NP_037185.1 | 3, 4 | 495 | ||
| GenBank mRNA | BC062409.1 | - | 2, 3 | 465 | 5623850 | |
| D17614.1 | - | 3, 4 | 495 | |||
| FQ218679.1 | - | 2, 3 | 255 | |||