siRNA Details
| Gene Symbol: Uqcr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170465.1 | NP_001163936.1 | 2 | 310 | ||
| GenBank mRNA | BC166935.1 | - | 2 | 294 | 6922239 | |
| CF978907.1 | - | 2 | 314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcr10| Gene Symbol: Uqcr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170465.1 | NP_001163936.1 | 2 | 310 | ||
| GenBank mRNA | BC166935.1 | - | 2 | 294 | 6922239 | |
| CF978907.1 | - | 2 | 314 | |||