siRNA Details
| Gene Symbol: Rnf11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245310.2 | XP_006245372.1 | 1 | 1490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf11l2| Gene Symbol: Rnf11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245310.2 | XP_006245372.1 | 1 | 1490 | ||