siRNA Details
| Gene Symbol: LOC102551267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766905.1 | XP_008765127.1 | 1 | 347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102551267| Gene Symbol: LOC102551267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766905.1 | XP_008765127.1 | 1 | 347 | ||