siRNA Details
Gene Symbol: LOC103693254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767450.1 | XP_008765672.1 | 1, 2 | 1366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC103693254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767450.1 | XP_008765672.1 | 1, 2 | 1366 |