siRNA Details
| Gene Symbol: EMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057571.1 | - | 2 | 2193 | ||
| AK222615.1 | - | 9 | 695 | |||
| AK290974.1 | - | 9 | 681 | |||
| BC020753.1 | - | 9 | 693 | 4809693 | ||
| BC021667.1 | - | 9 | 710 | 3682913 | ||
| CR457402.1 | - | 9 | 671 | |||
| D14659.1 | - | 9 | 677 | |||

EMC2
Genomic Map