siRNA Details
| Gene Symbol: LOC690435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047975.2 | NP_001041440.2 | 2 | 106 | ||
| GenBank mRNA | AY325131.1 | - | 2 | 106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690435| Gene Symbol: LOC690435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047975.2 | NP_001041440.2 | 2 | 106 | ||
| GenBank mRNA | AY325131.1 | - | 2 | 106 | ||