siRNA Details
| Gene Symbol: Ndufab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106294.1 | NP_001099764.1 | 2 | 279 | ||
| XM_006230188.1 | XP_006230250.1 | 2 | 311 | |||
| GenBank mRNA | FQ212307.1 | - | 2 | 278 | ||
| FQ212475.1 | - | 2 | 316 | |||
| FQ216942.1 | - | 2 | 256 | |||

Ndufab1
Genomic Map