siRNA Details
| Gene Symbol: LOC683897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109448.1 | NP_001102918.1 | 2, 3 | 558 | ||
| XM_006256581.2 | XP_006256643.1 | 3, 4 | 605 | |||
| GenBank mRNA | BC105890.1 | - | 1, 2 | 487 | 7306492 | |

LOC683897
Genomic Map