siRNA Details
| Gene Symbol: Bend3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256584.2 | XP_006256646.1 | 4 | 1539 | ||
| XM_008773024.1 | XP_008771246.1 | 3 | 1155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bend3| Gene Symbol: Bend3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256584.2 | XP_006256646.1 | 4 | 1539 | ||
| XM_008773024.1 | XP_008771246.1 | 3 | 1155 | |||