siRNA Details
| Gene Symbol: Cstf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131014.1 | NP_001124486.1 | 13 | 1696 | ||
| XM_008773440.1 | XP_008771662.1 | 15 | 1948 | |||
| GenBank mRNA | BC168251.1 | - | 13 | 1696 | 5599766 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cstf2| Gene Symbol: Cstf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131014.1 | NP_001124486.1 | 13 | 1696 | ||
| XM_008773440.1 | XP_008771662.1 | 15 | 1948 | |||
| GenBank mRNA | BC168251.1 | - | 13 | 1696 | 5599766 | |