siRNA Details
| Gene Symbol: Anapc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173983.1 | NP_001167454.1 | 3 | 355 | ||
| XM_006243681.2 | XP_006243743.1 | 3 | 654 | |||
| XM_006243682.2 | XP_006243744.1 | 3 | 1108 | |||
| XM_006243683.2 | XP_006243745.1 | 3 | 628 | |||

Anapc13
Genomic Map