siRNA Details
| Gene Symbol: Pwwp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246135.2 | XP_006246197.1 | 2 | 3006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pwwp2a| Gene Symbol: Pwwp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246135.2 | XP_006246197.1 | 2 | 3006 | ||