siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_012288.3 | NP_036420.1 | 7 | 744 | ||
| XM_011515005.2 | XP_011513307.1 | 7 | 619 | |||
| GenBank mRNA | AK291751.1 | - | 7 | 630 | ||
| AK311560.1 | - | 7 | 613 | |||
| AY927571.1 | - | LIT3572 AY927571 |
1 | 310 | ||
| BC028121.1 | - | 7 | 744 | 5169808 | ||
| D31762.1 | - | 7 | 670 | |||
| KU178488.1 | - | 7 | 595 | |||
| KU178489.1 | - | 6 | 531 | |||

TRAM2
Genomic Map