siRNA Details
| Gene Symbol: Flvcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769906.1 | XP_008768128.1 | 6 | 1361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flvcr1| Gene Symbol: Flvcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769906.1 | XP_008768128.1 | 6 | 1361 | ||