siRNA Details
| Gene Symbol: LOC681300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257539.2 | XP_006257601.1 | 1 | 108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC681300| Gene Symbol: LOC681300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257539.2 | XP_006257601.1 | 1 | 108 | ||