siRNA Details
| Gene Symbol: Krtcap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109674.1 | NP_001103144.1 | 4, 5 | 596 | ||
| GenBank mRNA | BC100633.1 | - | 4, 5 | 543 | 5622000 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtcap3| Gene Symbol: Krtcap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109674.1 | NP_001103144.1 | 4, 5 | 596 | ||
| GenBank mRNA | BC100633.1 | - | 4, 5 | 543 | 5622000 | |