siRNA Details
| Gene Symbol: Nup43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128191.1 | NP_001121663.1 | 1 | 73 | ||
| XM_006227622.2 | XP_006227684.1 | 1 | 332 | |||
| GenBank mRNA | BC158756.1 | - | 1 | 69 | 9103164 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup43| Gene Symbol: Nup43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128191.1 | NP_001121663.1 | 1 | 73 | ||
| XM_006227622.2 | XP_006227684.1 | 1 | 332 | |||
| GenBank mRNA | BC158756.1 | - | 1 | 69 | 9103164 | |