siRNA Details
Gene Symbol: LOC102548917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252956.2 | XP_006253018.1 | 4 | 675 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102548917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252956.2 | XP_006253018.1 | 4 | 675 |