siRNA Details
| Gene Symbol: Skint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135916.1 | NP_001129388.1 | 4 | 542 | ||
| GenBank mRNA | EU650232.1 | - | 4 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skint1| Gene Symbol: Skint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135916.1 | NP_001129388.1 | 4 | 542 | ||
| GenBank mRNA | EU650232.1 | - | 4 | 542 | ||