siRNA Details
| Gene Symbol: LOC100910720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771481.1 | XP_008769703.1 | 420 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100910720| Gene Symbol: LOC100910720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771481.1 | XP_008769703.1 | 420 | |||