siRNA Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233102.2 | XP_006233164.1 | 4 | 697 | ||
| XM_006233103.2 | XP_006233165.1 | 5 | 1656 | |||
| XM_008761434.1 | XP_008759656.1 | 6 | 1255 | |||
| XM_008761435.1 | XP_008759657.1 | 4 | 721 | |||

Fam19a3
Genomic Map