siRNA Details
| Gene Symbol: LRRTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109374.1 | NP_001102844.1 | 2 | 1230 | ||
| XM_006236700.2 | XP_006236762.1 | 2 | 1432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRTM1| Gene Symbol: LRRTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109374.1 | NP_001102844.1 | 2 | 1230 | ||
| XM_006236700.2 | XP_006236762.1 | 2 | 1432 | |||