siRNA Details
| Gene Symbol: Rp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195676.1 | NP_001182605.1 | 4 | 1197 | ||
| XM_008763558.1 | XP_008761780.1 | 5 | 1147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rp1| Gene Symbol: Rp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195676.1 | NP_001182605.1 | 4 | 1197 | ||
| XM_008763558.1 | XP_008761780.1 | 5 | 1147 | |||