siRNA Details
| Gene Symbol: LOC100910679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763462.1 | XP_008761684.1 | 2 | 151 | ||
| Gene Symbol: LOC502907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763434.1 | XP_008761656.1 | 3 | 818 | ||

LOC100910679
Genomic Map