siRNA Details
Gene Symbol: Nr2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113197.1 | NP_001106668.1 | 8 | 1075 | ||
GenBank mRNA | EU316216.1 | - | 8 | 1075 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113197.1 | NP_001106668.1 | 8 | 1075 | ||
GenBank mRNA | EU316216.1 | - | 8 | 1075 |