siRNA Details
| Gene Symbol: Ackr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243689.2 | XP_006243751.1 | 2 | 982 | ||
| XM_008766591.1 | XP_008764813.1 | 2 | 1352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ackr4| Gene Symbol: Ackr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243689.2 | XP_006243751.1 | 2 | 982 | ||
| XM_008766591.1 | XP_008764813.1 | 2 | 1352 | |||