siRNA Details
| Gene Symbol: Snx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044283.1 | NP_001037748.1 | 4 | 772 | ||
| GenBank mRNA | BC086341.1 | - | 4 | 536 | 7301779 | |
| BC107918.1 | - | 4 | 772 | 7458016 | ||
| FQ211490.1 | - | 4 | 477 | |||
| FQ234595.1 | - | 4 | 519 | |||

Snx3
Genomic Map