siRNA Details
| Gene Symbol: LOC100911148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229637.2 | XP_006229699.1 | 4 | 1728 | ||
| Gene Symbol: LOC690082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759620.1 | XP_008757842.1 | 2 | 2604 | ||

LOC100911148
Genomic Map