siRNA Details
| Gene Symbol: Trabd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001072500.4 | XP_001072500.2 | 3 | 884 | ||
| XM_006238601.2 | XP_006238663.1 | 3 | 884 | |||
| XM_006238603.2 | XP_006238665.1 | 3 | 884 | |||
| XM_006238605.2 | XP_006238667.1 | 3 | 884 | |||
| XM_006238606.2 | XP_006238668.1 | 3 | 884 | |||
| XM_008763972.1 | XP_008762194.1 | 3 | 884 | |||
| XM_008763973.1 | XP_008762195.1 | 3 | 884 | |||
| XM_008763974.1 | XP_008762196.1 | 3 | 884 | |||

Trabd2b
Genomic Map