siRNA Details
| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134985.1 | NP_001128457.1 | 9 | 1118 | ||
| GenBank mRNA | BC169033.1 | - | 9 | 1118 | 7114893 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcor3| Gene Symbol: Rcor3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134985.1 | NP_001128457.1 | 9 | 1118 | ||
| GenBank mRNA | BC169033.1 | - | 9 | 1118 | 7114893 | |