siRNA Details
| Gene Symbol: LOC102557071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769021.1 | XP_008767243.1 | 1 | 20 | ||
| XM_008769022.1 | XP_008767244.1 | 1 | 20 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102557071| Gene Symbol: LOC102557071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769021.1 | XP_008767243.1 | 1 | 20 | ||
| XM_008769022.1 | XP_008767244.1 | 1 | 20 | |||