siRNA Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247890.2 | XP_006247952.1 | 7, 8 | 1676 | ||
| GenBank mRNA | BC168919.1 | - | 6, 7 | 1144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxk2| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247890.2 | XP_006247952.1 | 7, 8 | 1676 | ||
| GenBank mRNA | BC168919.1 | - | 6, 7 | 1144 | ||