siRNA Details
| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001072378.4 | XP_001072378.2 | 3, 4 | 1549 | ||
| XM_006236344.2 | XP_006236406.1 | 3, 4 | 1550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrg2| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001072378.4 | XP_001072378.2 | 3, 4 | 1549 | ||
| XM_006236344.2 | XP_006236406.1 | 3, 4 | 1550 | |||