siRNA Details
| Gene Symbol: Chchd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749892.3 | XP_003749940.1 | 3 | 468 | ||
| XM_003749893.2 | XP_003749941.1 | 3 | 307 | |||
| XM_008763563.1 | XP_008761785.1 | 3 | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chchd7| Gene Symbol: Chchd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749892.3 | XP_003749940.1 | 3 | 468 | ||
| XM_003749893.2 | XP_003749941.1 | 3 | 307 | |||
| XM_008763563.1 | XP_008761785.1 | 3 | 486 | |||