siRNA Details
| Gene Symbol: Fam64a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113781.2 | NP_001107253.2 | 2 | 100 | ||
| XM_006246759.2 | XP_006246821.1 | 2 | 210 | |||
| XM_006246760.2 | XP_006246822.1 | 2 | 186 | |||
| GenBank mRNA | BC158692.1 | - | 2 | 100 | 9022735 | |
| Gene Symbol: LOC100912261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766825.1 | XP_008765047.1 | 1 | 61 | ||

Fam64a
Genomic Map