siRNA Details
| Gene Symbol: Syt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751296.3 | XP_003751344.1 | 3 | 435 | ||
| XM_003751297.3 | XP_003751345.1 | 3 | 487 | |||
| XM_008769895.1 | XP_008768117.1 | 1 | 69 | |||
| XM_008769896.1 | XP_008768118.1 | 1 | 69 | |||
| XM_008769897.1 | XP_008768119.1 | 1 | 69 | |||

Syt14
Genomic Map