siRNA Details
Gene Symbol: Magohb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191105.1 | NP_001178034.1 | 3 | 206 | ||
XM_008763423.1 | XP_008761645.1 | 3 | 1482 | |||
XM_008763424.1 | XP_008761646.1 | 4 | 351 |

More Information
Other Species Targeted

Gene Ontology Categories:
