siRNA Details
| Gene Symbol: ADGRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001702.2 | NP_001693.2 | 27 | 4288 | ||
| XM_011517199.2 | XP_011515501.1 | 28 | 4726 | |||
| XM_017013691.1 | XP_016869180.1 | 30 | 4960 | |||
| XM_017013692.1 | XP_016869181.1 | 30 | 4960 | |||
| XM_017013693.1 | XP_016869182.1 | 30 | 4951 | |||
| XM_017013694.1 | XP_016869183.1 | 29 | 4861 | |||
| XM_017013695.1 | XP_016869184.1 | 29 | 4840 | |||
| XM_017013696.1 | XP_016869185.1 | 29 | 4825 | |||
| XM_017013697.1 | XP_016869186.1 | 29 | 4795 | |||
| GenBank mRNA | AB005297.1 | - | 27 | 4288 | ||
More Information
Set Membership: |
|

ADGRB1
Genomic Map