siRNA Details
| Gene Symbol: LOC679989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773460.1 | XP_008771682.1 | 7 | 969 | ||
| XM_008773461.1 | XP_008771683.1 | 5 | 659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC679989| Gene Symbol: LOC679989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773460.1 | XP_008771682.1 | 7 | 969 | ||
| XM_008773461.1 | XP_008771683.1 | 5 | 659 | |||