siRNA Details
Gene Symbol: LOC689959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109559.1 | NP_001103029.1 | 4 | 334 | ||
GenBank mRNA | FQ220607.1 | - | 4 | 251 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC689959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109559.1 | NP_001103029.1 | 4 | 334 | ||
GenBank mRNA | FQ220607.1 | - | 4 | 251 |