siRNA Details
| Gene Symbol: Kcmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128192.1 | NP_001121664.1 | 1 | 188 | ||
| GenBank mRNA | BC167075.1 | - | 1 | 188 | 9093013 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019715, BC043330, BC029325, AK007070, AK142436.

Kcmf1
Genomic Map